4-chlorotetrahydropyran, 96%
4-chlorotetrahydropyran, 96%
4-chlorotetrahydropyran, 96%
Thermo Scientific Chemicals

4-chlorotetrahydropyran, 96%

CAS: 1768-64-5 | C5H9ClO | 120.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 434270050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1768-64-5
IUPAC Name4-chlorooxane
Molecular FormulaC5H9ClO
InChI KeyDHRSKOBIDIDMJZ-UHFFFAOYSA-N
SMILESClC1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Colorless to dark yellow
GC>=95.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only