BioProbes™ Journal of Cell Biology Applications—Subscribe today »
Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits
Product Image
Invitrogen™

Streptavidin - Cy3

Streptavidin - Cy3
Have Questions?
Catalog NumberQuantity
4343151 mL
Catalog number 434315
Price (USD)
283.00
Each
Quantity:
1 mL
Price (USD)
283.00
Each

Reactivity: CY3-SA reacts with biotin.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Concentration1.5 mg/mL
Product TypeStreptavidin Conjugate (fluorescent)
Quantity1 mL
Shipping ConditionWet Ice
ConjugateCy3
FormLiquid
Product LineCy
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C).