BioProbes™ Journal of Cell Biology Applications—Subscribe today »
Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits
Streptavidin - AP
Invitrogen™

Streptavidin - AP

Streptavidin - AP
Have Questions?
Catalog NumberQuantity
4343223 mg
Catalog number 434322
Price (USD)
313.00
Each
Quantity:
3 mg
Price (USD)
313.00
Each

Reactivity: AP-SA reacts with biotin.

Applications: Immunohistochemistry, Western blotting, and ELISA

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Concentration1.5 mg/mL
Label or DyeAP (Alkaline Phosphatase)
Product TypeStreptavidin Conjugate (fluorescent)
Quantity3 mg
Shipping ConditionWet Ice
ConjugateAP
FormLiquid
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C).

Citations & References (1)

Citations & References
Abstract
Vitronectin gene expression in vivo. Evidence for extrahepatic synthesis and acute phase regulation.
Authors:Seiffert D, Crain K, Wagner NV, Loskutoff DJ
Journal:J Biol Chem
PubMed ID:7519600
'A competitive polymerase chain reaction (PCR) assay was developed to quantitate vitronectin (Vn) mRNA in murine tissues using a synthetic RNA as an external standard. Although the liver contained the highest concentration of Vn mRNA, significant levels were also detected in the brain (25-fold less) and in adipose tissue, heart, ... More