MicroAmp™ Optical 384-Well Reaction Plate - FAQs
MicroAmp™ Optical 384-Well Reaction Plate - FAQs
View additional product information for MicroAmp™ Optical 384-Well Reaction Plate - FAQs (4343370)
1 product FAQs found
Are the MicroAmp Optical 96-Well Reaction Plates and MicroAmp Optical 384 -Well Reaction Plates autoclavable?
Our microplates are certified to be DNA, RNase and PCR inhibitor free as long as they are kept in the original packaging bag. They are not meant to be autoclaved.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.