Why have the amplicon lengths for the Quantifiler Duo DNA Quantification Kit been increased compared to the previous Quantifiler Y and Quantifiler Human DNA Quantification Kits?
The sizes of the amplicons were chosen to improve the estimation of DNA quantity, thereby delivering a more realistic estimate of the amount of DNA present in degraded samples and providing better correlation between the quantitation values and the performance of the AmpFℓSTR kits.
What does the "HP" in the Quantifiler HP DNA Quantification Kit name signify?
The "HP" stands for "Human Plus" because this kit is intended as an upgrade to the Quantifiler Human DNA Quantification Kit. Like the Quantifiler Human kit, the new Quantifiler HP kit detects and quantifies human DNA (without a male target), but the new kit also includes a large autosomal (LA) target to enable degradation index functionality and more comprehensive sample quality assessment. It also features increased sensitivity and robustness to better correlate with Applied Biosystems STR chemistries such as those in the Identifiler Plus, NGM SElect, and GlobalFiler PCR Amplification Kits.