BioProbes™ Journal of Cell Biology Applications—Subscribe today »
Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits
Avidin, AP conjugate
Invitrogen™

Avidin, AP conjugate

Avidin, AP conjugate
Have Questions?
Catalog NumberQuantity
4344222 mL
Catalog number 434422
Price (USD)
538.65
Online exclusive
718.00
Save 179.35
Each
Quantity:
2 mL
Price (USD)
538.65
Online exclusive
718.00
Save 179.35
Each

Reactivity: AP-Avidin reacts with biotin.

Applications: Immunohistochemistry, ELISA

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Label or DyeAP (Alkaline Phosphatase)
Quantity2 mL
Shipping ConditionWet Ice
ConjugateAP
FormLiquid
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C).

Citations & References (2)

Citations & References
Abstract
Humoral response to a carboxyl-terminal region of the merozoite surface protein-1 plays a predominant role in controlling blood-stage infection in rodent malaria.
Authors:Daly TM, Long CA
Journal:J Immunol
PubMed ID:7602100
'The developmental stages of malaria parasites that infect E are responsible for the morbidity and mortality associated with this disease. One of the leading candidates for a blood-stage vaccine against malaria is a surface protein of merozoites, the infectious stages for E, designated merozoite surface protein-1 (MSP-1). The rodent malarial ... More
Transitions to cigarette smoking during adolescence.
Authors:Fergusson DM, Horwood LJ
Journal:Addict Behav
PubMed ID:8712060
The process of transition from nonsmoking to regular weekly smoking during a period from 10 to 16 years was examined using data gathered during the course of a longitudinal study of 957 New Zealand adolescents. These data were analyzed using a latent Markov model to estimate both rates of transition ... More