Dilithium tetrabromonickelate(II), 0.5M solution in THF, AcroSeal™
Dilithium tetrabromonickelate(II), 0.5M solution in THF, AcroSeal™
Dilithium tetrabromonickelate(II), 0.5M solution in THF, AcroSeal™
Dilithium tetrabromonickelate(II), 0.5M solution in THF, AcroSeal™
Thermo Scientific Chemicals

Dilithium tetrabromonickelate(II), 0.5M solution in THF, AcroSeal™

CAS: 13826-95-4 | Br4Li2Ni | 392.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 434541000
Price (EUR)
43,70
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (EUR)
43,70
Each
Chemical Identifiers
CAS13826-95-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Blue-green to brown
Titration Argentometric0.45 to 0.55 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only