1-Octanol, 99%, anhydrous, AcroSeal™
1-Octanol, 99%, anhydrous, AcroSeal™
1-Octanol, 99%, anhydrous, AcroSeal™
1-Octanol, 99%, anhydrous, AcroSeal™
Thermo Scientific Chemicals

1-Octanol, 99%, anhydrous, AcroSeal™

CAS: 111-87-5 | C8H18O | 130.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number 434581000
Price (USD)
37.65
Special offer
Online exclusive
Ends: 28-Dec-2026
43.90
Save 6.25
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
37.65
Special offer
Online exclusive
Ends: 28-Dec-2026
43.90
Save 6.25
Each
Chemical Identifiers
CAS111-87-5
IUPAC Nameoctan-1-ol
Molecular FormulaC8H18O
InChI KeyKBPLFHHGFOOTCA-UHFFFAOYSA-N
SMILESCCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.005 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
Residue after evaporation=<0.004 %
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only