n-Dodecane, 99%, anhydrous, AcroSeal™
n-Dodecane, 99%, anhydrous, AcroSeal™
n-Dodecane, 99%, anhydrous, AcroSeal™
n-Dodecane, 99%, anhydrous, AcroSeal™
Thermo Scientific Chemicals

n-Dodecane, 99%, anhydrous, AcroSeal™

CAS: 112-40-3 | C12H26 | 170.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number 434591000
Price (USD)
137.65
Special offer
Online exclusive
Ends: 30-Jun-2026
162.00
Save 24.35 (15%)
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
137.65
Special offer
Online exclusive
Ends: 30-Jun-2026
162.00
Save 24.35 (15%)
Each
Chemical Identifiers
CAS112-40-3
IUPAC Namedodecane
Molecular FormulaC12H26
InChI KeySNRUBQQJIBEYMU-UHFFFAOYSA-N
SMILESCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Infrared spectrumConforms
Appearance (Color)Clear colorless
Water=<0.005 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only