TaqMan™ Array, Format 64 - FAQs
TaqMan™ Array, Format 64 - FAQs
View additional product information for TaqMan™ Array, Format 64 - FAQs (4346800)
1 product FAQs found
What are the storage recommendations for TaqMan Array Cards (TAC)?
We recommend storing TaqMan Array Cards (TAC) at 2-8 degrees C. TaqMan Array Cards (TAC) are stable for 2 years from the manufacturing date. The expiration date is listed on the packaging.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.