Kaempferol, 97%
Kaempferol, 97%
Kaempferol, 97%
Thermo Scientific Chemicals

Kaempferol, 97%

CAS: 520-18-3 | C15H10O6 | 286.24 g/mol
Have Questions?
Catalog NumberQuantityPackaging
434731000100 mgGlass bottle
Catalog number 434731000
Price (USD)
335.00
Each
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
335.00
Each
Chemical Identifiers
CAS520-18-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Appearance (Color)Yellow
Water=<10 % (K.F.)
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only