MicroSEQ™ 500 16S rDNA PCR Kit, with protocol - FAQs
MicroSEQ™ 500 16S rDNA PCR Kit, with protocol - FAQs
View additional product information for MicroSEQ™ 500 16S rDNA PCR Kit, with protocol - FAQs (4348229)
1 product FAQs found
Is there a DNA extraction kit that you recommend using along with the MicroSeq 500 kit?
Yes, DNA extraction is greatly simplified by using the Applied Biosystems PrepMan Ultra Sample Preparation Reagent (Cat. No. 4322537), which can be used for all types of bacteria.