1-BOC-4-chloropiperidine, 97%
1-BOC-4-chloropiperidine, 97%
1-BOC-4-chloropiperidine, 97%
Thermo Scientific Chemicals

1-BOC-4-chloropiperidine, 97%

CAS: 154874-94-9 | C10H18ClNO2 | 219.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 435010010
Price (USD)
134.65
Special offer
Online exclusive
Ends: 28-Dec-2026
158.00
Save 23.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
134.65
Special offer
Online exclusive
Ends: 28-Dec-2026
158.00
Save 23.35
Each
Chemical Identifiers
CAS154874-94-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Infrared spectrumConforms
Appearance (Form)Liquid or low melting solid
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only