TAQMAN 384 ARRAY INSTALL RNASE - FAQs
TAQMAN 384 ARRAY INSTALL RNASE - FAQs
View additional product information for TAQMAN 384 ARRAY INSTALL RNASE - FAQs (4351469)
1 product FAQs found
Can I reuse my calibration plates on the 7900HT Fast Real-Time PCR System?
The calibration plates can be stored and reused three times for up to 12 months after you first open them, so make sure to return them to their original packaging and return them to -20 degrees C storage until the next use. (Note: Due to the small volumes, calibration array cards cannot be reused. Please discard them after use.) If needed, you can make your own background plate using deionized water.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.