3-Aminophenylboronic acid hydrochloride, 98%
3-Aminophenylboronic acid hydrochloride, 98%
3-Aminophenylboronic acid hydrochloride, 98%
Thermo Scientific Chemicals

3-Aminophenylboronic acid hydrochloride, 98%

CAS: 85006-23-1 | C6H9BClNO2 | 173.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 435530010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS85006-23-1
IUPAC Name(3-aminophenyl)boronic acid hydrochloride
Molecular FormulaC6H9BClNO2
InChI KeyQBMHZZHJIBUPOX-UHFFFAOYSA-N
SMILESCl.NC1=CC=CC(=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Elemental Analysis7.88 to 8.28 % N
HPLC>=98.0 %
Appearance (Color)Off-white to grey
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only