N-BOC-Hexahydro-1H-azepin-4-one, 97%
N-BOC-Hexahydro-1H-azepin-4-one, 97%
N-BOC-Hexahydro-1H-azepin-4-one, 97%
Thermo Scientific Chemicals

N-BOC-Hexahydro-1H-azepin-4-one, 97%

CAS: 188975-88-4 | C11H19NO3 | 213.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 435540010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS188975-88-4
IUPAC Nametert-butyl 4-oxoazepane-1-carboxylate
Molecular FormulaC11H19NO3
InChI KeyPMLBUVZPRKXMOX-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)N1CCCC(=O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Form)Liquid or low melting solid
Appearance (Color)Colorless to yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only