MicroAmp™ Fast 8-Tube Strip, 0.1 mL - FAQs
MicroAmp™ Fast 8-Tube Strip, 0.1 mL - FAQs
View additional product information for MicroAmp™ Fast 8-Tube Strip, 0.1 mL - FAQs (4358293)
1 product FAQs found
Can I run strip tubes on the ViiA 7 instrument?
Yes. Make sure to use the appropriate matching parts. For the 96-well standard block, use the standard 96-well plate adapter (Cat. No. 4459845), the retainer tray for (0.2 mL) tubes, and Optical 8-Tube Strips and Caps (Cat. No. 4316567 and 4323032). For the 96-well Fast block, use the Fast 96-well plate adapter (Cat. No. 4462078), the retainer tray (Cat. No. 4379983), and Fast (0.1 mL) Optical 8-tube Caps and Strips (Cat. Nos. 4323032 and 4358293).
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.