Methyl 2-(hydroxymethyl)acrylate, 85%, stabilized with 0.1 wt.% MEHQ
Methyl 2-(hydroxymethyl)acrylate, 85%, stabilized with 0.1 wt.% MEHQ
Methyl 2-(hydroxymethyl)acrylate, 85%, stabilized with 0.1 wt.% MEHQ
Thermo Scientific Chemicals

Methyl 2-(hydroxymethyl)acrylate, 85%, stabilized with 0.1 wt.% MEHQ

CAS: 15484-46-5 | C5H8O3 | 116.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 435900050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS15484-46-5
SpecificationsSpecification SheetSpecification Sheet
GC>=84.0 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only