Plates and Modules with Covalent Binding Surfaces
Plates and Modules with Covalent Binding Surfaces
Plates and Modules with Covalent Binding Surfaces
Plates and Modules with Covalent Binding Surfaces
Thermo Scientific™

Plates and Modules with Covalent Binding Surfaces

Reduce the amount of reagents used in the lab by using these plates and modules with covalent binding surfaces.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorVolume (Metric) WorkingProduct TypeSurface Treatment
436007White100 μLSolid plate 96-WellCovalent Binding
436006Clear100 μLMicroplateAmine
436008Black100 μLMicroplateAmine
436009Clear50 μLSolid plate 384-WellAmine
436013Clear100 μLMicroplateAmine
436023Clear100 μLMicroplateAmine
478042Clear100 μLSolid Module F8 (x12) in FrameCovalent Binding
Catalog number 436007
Price (USD)
593.29
Online exclusive
911.23
Save 317.94
Case of 30
Color:
White
Volume (Metric) Working:
100 μL
Product Type:
Solid plate 96-Well
Surface Treatment:
Covalent Binding
Price (USD)
593.29
Online exclusive
911.23
Save 317.94
Case of 30

Reduce the amount of reagents used in the lab with Thermo Scientific™ Plates and Modules with Covalent Binding Surfaces.

Covalent Surfaces (Thermo Scientific™ CovaLink™ and Immobilizer Amino)

  • Minimal leaching
  • Withstands vigorous washing
  • Coating with lower amounts of reagent may be possible
  • Control of orientation

Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.

WARNING: Cancer – www.P65Warnings.ca.gov
Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.
Specifications
Binding TypeCovalent
Detection MethodChemiluminescence
FormatStandard
No. per Case30
No. per Pack5
Packaging30 Cases
Plate BlockingUnspecified
Product TypeSolid plate 96-Well
Target MoleculePeptides
Total Volume (Metric) Well400 μL
Volume (Metric) Well400 μL
Volume (Metric) Working100 μL
Array8 x 12
ColorWhite
MaterialPolystyrene
No. of Wells96
SterilityNon-sterile
Surface TreatmentCovalent Binding
Well ShapeRound, Flat Bottom (F-well)
Unit SizeCase of 30

Frequently asked questions (FAQs)

Which type of Immobilizer Amino plates and strips should I use for the different detection methods?

We recommend using transparent polystyrene plates and strips for colorimetric assays, white opaque polystyrene plates and strips for bio and chemi-luminescence assays, and black opaque polystyrene plates and strips for fluorescence assays.

Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with all Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates?

Our Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules were designed to provide a positive seal for flat and round bottom wells of our Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates.
8-Well Strip Caps for Immuno Standard Modules are not compatible with C or V bottom wells of the Nunc Microwell 96-Well Microplates and Nunc Immuno 96-Well Plates.

For Nunc Immuno plates and modules, what are the advantages of one well geometry type over another? Which types should I use for various applications?

The following list describes the geometries of wells available for Nunc Immuno-plates and modules:
- Flat bottom (F): Allows maximum transmission of light. These plates can be read on a monochromatic reader.
- Round bottom (U): This geometry optimizes washing and coating. These plates can be read using a dual wavelength reader.
- "C" bottom (C): This design of the well is a combination of both flat and round bottoms. Basically, it is a flat bottomed well with curved edges at the bottom. These plates also can be read using a monochromatic reader combining optimal reading and washing.
- StarWell: These wells have a modified "C" shape geometry with eight fins strategically placed at the bottom. This feature increases surface area, allowing more molecules to become immobilized which reduces incubation times.

What advantages do your covalent binding surfaces offer over passive surfaces?

While adsorption of biomolecules to our passive surfaces is more electrostatic in nature, our CovaLink and Immobilizer Amino surfaces present an option for true covalent bonding of molecules with specific free residues. CovaLink possesses a secondary amine structure, which will covalently bond to biomolecules with a free carboxyl or phosphate group. This allows for a much more robust and specific coating, which will often tolerate more rigorous washing steps and eliminate the need for blocking.