Plates and Modules with Covalent Binding Surfaces, 400μL, clear, F8 module and frame, amino - FAQs
Plates and Modules with Covalent Binding Surfaces, 400μL, clear, F8 module and frame, amino - FAQs
View additional product information for Plates and Modules with Covalent Binding Surfaces - FAQs (436009, 436008, 436007, 478042, 436013, 436023, 436006)
3 product FAQs found
Which type of Immobilizer Amino plates and strips should I use for the different detection methods?
We recommend using transparent polystyrene plates and strips for colorimetric assays, white opaque polystyrene plates and strips for bio and chemi-luminescence assays, and black opaque polystyrene plates and strips for fluorescence assays.
Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with all Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates?
Our Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules were designed to provide a positive seal for flat and round bottom wells of our Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates.
8-Well Strip Caps for Immuno Standard Modules are not compatible with C or V bottom wells of the Nunc Microwell 96-Well Microplates and Nunc Immuno 96-Well Plates.
What advantages do your covalent binding surfaces offer over passive surfaces?
While adsorption of biomolecules to our passive surfaces is more electrostatic in nature, our CovaLink and Immobilizer Amino surfaces present an option for true covalent bonding of molecules with specific free residues. CovaLink possesses a secondary amine structure, which will covalently bond to biomolecules with a free carboxyl or phosphate group. This allows for a much more robust and specific coating, which will often tolerate more rigorous washing steps and eliminate the need for blocking.