Plates and Modules with Affinity Binding Surfaces
Plates and Modules with Affinity Binding Surfaces
Plates and Modules with Affinity Binding Surfaces
Plates and Modules with Affinity Binding Surfaces
Thermo Scientific™

Plates and Modules with Affinity Binding Surfaces

Help reduce variability in molecular orientation with these plates and modules with affinity binding surfaces.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorProduct TypeSurface TreatmentTotal Volume (Metric) Well
436017ClearMicroplateStreptavidin120 μL
436014ClearMicroplateStreptavidin400 μL
436015WhiteMicroplateStreptavidin400 μL
436016BlackMicroplateStreptavidin400 μL
436020ClearSolid Module F8 (x12) in FrameStreptavidin400 μL
436022ClearLock-Well Module C8 (x12) in FrameStreptavidin350 μL
436033WhiteMicroplateGlutathione400 μL
95029263ClearMicroplateStreptavidin400 μL
95029293ClearBreakable Module F8 (x12) in FrameStreptavidin400 μL
Catalog number 436017
Price (USD)
909.59
Online exclusive
1,060.88
Save 151.29
Case of 15
Color:
Clear
Product Type:
Microplate
Surface Treatment:
Streptavidin
Total Volume (Metric) Well:
120 μL
Price (USD)
909.59
Online exclusive
1,060.88
Save 151.29
Case of 15

Help reduce variability in molecular orientation with Thermo Scientific™ Plates and Modules with Affinity Binding Surfaces.

Affinity Capture Surfaces (Passive Streptavidin and Immobilizer Streptavidin, Nickel Chelate and Glutathione, BioBind):

  • Highly specific binding surfaces reduce variability in molecular orientation.
  • Immobilizer surfaces improve signal to noise ratios.
  • Streptavidin biotin interaction can be exploited to immobilize a wide range of biomolecules.
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Binding TypeAffinity
Detection MethodColorimetric
FormatStandard
LidWithout Lid
No. per Case15
No. per Pack1
Packaging15 Cases
Plate BlockingUnspecified
Product TypeMicroplate
Quantity15/Cs.
Target MoleculeBiotinylated
Total Volume (Metric) Well120 μL
Volume (Metric) Well120 μL
Volume (Metric) Working50 μL
ColorClear
MaterialPolystyrene
No. of Wells384
SterilityNon-sterile
Surface TreatmentStreptavidin
Well ShapeSquare, Flat Bottom (F-well)
Unit SizeCase of 15

Frequently asked questions (FAQs)

Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with all Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates?

Our Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules were designed to provide a positive seal for flat and round bottom wells of our Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates.
8-Well Strip Caps for Immuno Standard Modules are not compatible with C or V bottom wells of the Nunc Microwell 96-Well Microplates and Nunc Immuno 96-Well Plates.

For Nunc Immuno plates and modules, what are the advantages of one well geometry type over another? Which types should I use for various applications?

The following list describes the geometries of wells available for Nunc Immuno-plates and modules:
- Flat bottom (F): Allows maximum transmission of light. These plates can be read on a monochromatic reader.
- Round bottom (U): This geometry optimizes washing and coating. These plates can be read using a dual wavelength reader.
- "C" bottom (C): This design of the well is a combination of both flat and round bottoms. Basically, it is a flat bottomed well with curved edges at the bottom. These plates also can be read using a monochromatic reader combining optimal reading and washing.
- StarWell: These wells have a modified "C" shape geometry with eight fins strategically placed at the bottom. This feature increases surface area, allowing more molecules to become immobilized which reduces incubation times.

Why am I getting a high background signal when using Nunc Immobilizer Streptavidin Plates/Strips?

High background signal could be due to inadequate washing. Make sure to pre-wash the Nunc Immobilizer Streptavidin Plates/Strips and increase the number of washing steps and the time between washes. Another possible cause is too high a concentration of the detection components.

How long will my Nunc Immobilizer Streptavidin Plates/Strips remain stable?

Our Nunc Immobilizer Streptavidin Plates/Strips are extremely stable over time, if stored at room temperature (20 degrees C to 25 degrees C). If kept in the unopened foil packaging, this product should have an 18 month shelf life after the date of manufacturing.

How long should I incubate the biotinylated and target molecule with the Nunc Immobilizer Streptavidin surface?

Coupling of the target molecule to the streptavidin proceeds very rapidly. Results show that incubation with gentle agitation for 2 hr at room temperature is typically sufficient to saturate all available active site.