Plates and Modules with Covalent Binding Surfaces
Plates and Modules with Covalent Binding Surfaces
Plates and Modules with Covalent Binding Surfaces
Plates and Modules with Covalent Binding Surfaces
Thermo Scientific™

Plates and Modules with Covalent Binding Surfaces

Reduce the amount of reagents used in the lab by using these plates and modules with covalent binding surfaces.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorVolume (Metric) WorkingProduct TypeSurface Treatment
436023Clear100 μLMicroplateAmine
436006Clear100 μLMicroplateAmine
436007White100 μLSolid plate 96-WellCovalent Binding
436008Black100 μLMicroplateAmine
436009Clear50 μLSolid plate 384-WellAmine
436013Clear100 μLMicroplateAmine
478042Clear100 μLSolid Module F8 (x12) in FrameCovalent Binding
Catalog number 436023
Price (USD)
944.54
Online exclusive
1,102.28
Save 157.74
Case of 30
Color:
Clear
Volume (Metric) Working:
100 μL
Product Type:
Microplate
Surface Treatment:
Amine
Price (USD)
944.54
Online exclusive
1,102.28
Save 157.74
Case of 30

Reduce the amount of reagents used in the lab with Thermo Scientific™ Plates and Modules with Covalent Binding Surfaces.

Covalent Surfaces (Thermo Scientific™ CovaLink™ and Immobilizer Amino)

  • Minimal leaching
  • Withstands vigorous washing
  • Coating with lower amounts of reagent may be possible
  • Control of orientation

Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.
Specifications
Binding TypeCovalent
BreakapartBreakable Strips
Detection MethodColorimetric
No. per Case30
No. per Pack5
Packaging30 Cases
Plate BlockingUnspecified
Product TypeMicroplate
Target MoleculePeptides
Total Volume (Metric) Well350 μL
Volume (Metric) Well350 μL
Volume (Metric) Working100 μL
ColorClear
MaterialPolystyrene
SterilityNon-sterile
Surface TreatmentAmine
Well ShapeRound, C Bottom
Unit SizeCase of 30

Frequently asked questions (FAQs)

Which type of Immobilizer Amino plates and strips should I use for the different detection methods?

We recommend using transparent polystyrene plates and strips for colorimetric assays, white opaque polystyrene plates and strips for bio and chemi-luminescence assays, and black opaque polystyrene plates and strips for fluorescence assays.

For Nunc Immuno plates and modules, what are the advantages of one well geometry type over another? Which types should I use for various applications?

The following list describes the geometries of wells available for Nunc Immuno-plates and modules:
- Flat bottom (F): Allows maximum transmission of light. These plates can be read on a monochromatic reader.
- Round bottom (U): This geometry optimizes washing and coating. These plates can be read using a dual wavelength reader.
- "C" bottom (C): This design of the well is a combination of both flat and round bottoms. Basically, it is a flat bottomed well with curved edges at the bottom. These plates also can be read using a monochromatic reader combining optimal reading and washing.
- StarWell: These wells have a modified "C" shape geometry with eight fins strategically placed at the bottom. This feature increases surface area, allowing more molecules to become immobilized which reduces incubation times.

What advantages do your covalent binding surfaces offer over passive surfaces?

While adsorption of biomolecules to our passive surfaces is more electrostatic in nature, our CovaLink and Immobilizer Amino surfaces present an option for true covalent bonding of molecules with specific free residues. CovaLink possesses a secondary amine structure, which will covalently bond to biomolecules with a free carboxyl or phosphate group. This allows for a much more robust and specific coating, which will often tolerate more rigorous washing steps and eliminate the need for blocking.