Plates and Modules with Affinity Binding Surfaces, 400μL, solid plate, clear, glutathione - FAQs
Plates and Modules with Affinity Binding Surfaces, 400μL, solid plate, clear, glutathione - FAQs
View additional product information for Plates and Modules with Affinity Binding Surfaces - FAQs (436032)
5 product FAQs found
What is the detection limit of proteins with Immobilizer Glutathione plates?
The background is extremely low when using the Immobilizer Glutathione plates. This results in a high signal-to-noise ratio and a low detection limit which in general means that the detection limit (OD 0.5) is 3 ng per well (100 µL).
How long are Immobilizer Glutathione plates stable?
The Immobilizer Glutathione plates are extremely stable over long periods of time, if stored at room temperature (20-25 degrees C). We guarantee 12 mths of shelf life in unopened foil package after the date of shipment.
With Immobilizer Glutathione plates, why should I use Tween-20 surfactant in the washing step and in the buffers containing the antibodies?
We recommend using 0.05% Tween-20 surfactant (as the non-ionic detergent) to improve the signal-to-noise ratio of the assay and thus decrease the background.
Should I optimize the concentration of the purified GST or GST-tagged protein to be used with Immobilizer Glutathione plates?
Yes. We recommend using a solution of the purified GST or GST-tagged protein diluted in Phosphate Buffered Saline (PBS), pH 7.2 in a concentration ranging from 0.01-1 µg/mL. During the coupling, non-ionic detergents like Tween-20 surfactant should not be present, as these will suppress the coupling.
Which type of molecules can I use with Immobilizer Glutathione plates?
The Immobilizer Glutathione plates are designed for optimal detection of glutathione-S-transferase (GST)-tagged proteins or purified GST.