Poly(DL-lactide-co-glycolide), 50:50 lactide glycolide, M.N. 5,000 to 10,000 Da
Poly(DL-lactide-co-glycolide), 50:50 lactide glycolide, M.N. 5,000 to 10,000 Da
Poly(DL-lactide-co-glycolide), 50:50 lactide glycolide, M.N. 5,000 to 10,000 Da
Thermo Scientific Chemicals

Poly(DL-lactide-co-glycolide), 50:50 lactide glycolide, M.N. 5,000 to 10,000 Da

CAS: 26780-50-7 | (C3H4O2)x(C2H2O2)y | NaN g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 436200010
Price (USD)
126.65
Special offer
Online exclusive
Ends: 28-Dec-2026
149.00
Save 22.35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
126.65
Special offer
Online exclusive
Ends: 28-Dec-2026
149.00
Save 22.35
Each
Chemical Identifiers
CAS26780-50-7
Molecular Formula(C3H4O2)x(C2H2O2)y
SMILESCC(-*)C(=O)OCC(=O)O-*
MDL NumberMFCD00131930
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Frequently asked questions (FAQs)

Is Poly(DL-lactide-co-glycolide), 50:50 lactide glycolide, M.N. 5,000 to 10,000 Da acid-terminated or ester-terminated?

Our Poly(DL-lactide-co-glycolide), 50:50 lactide glycolide, M.N. 5,000 to 10,000 Da is acid-terminated.