Primer Express™ Software v3.0.1 License
Applied Biosystems™
Primer Express™ Software v3.0.1 License
Primer Express Software v3.0.1 allows you to design your own primers and probes using TaqMan and SYBR Green I dyeRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
43639935 users
43639911 user
Catalog number 4363993
Price (MXN)
-
Quantity:
5 users
Primer Express Software v3.0.1 allows you to design your own primers and probes using TaqMan and SYBR Green I dye chemistries for gene quantitation and allelic discrimination (SNP) real-time PCR applications. Developed specifically for use with our StepOne, StepOnePlus, 7300, 7500, 7500 Fast, 7900HT, ViiA 7, and QuantStudio real-time PCR systems, Primer Express Software provides customized application-specific documents for absolute⁄relative quantitation and allelic discrimination.
Download Primer Express Software v3.0.1 (license key required) >
View Release Notes detailing enhancements in v3.0.1 >
• Enables individual oligonucleotide design for real-time PCR applications.
• Supports assays based on TaqMan and SYBR Green I dye chemistries.
• Provides design flexibility, ease-of-use, and requires minimal optimization.
• Provides robust assay performance when used in accordance with Assay Development Guidelines.
Easy-to-Learn Software
Primer Express Software v3.0.1 is flexible, easy to learn and to use, and requires minimal optimization. It includes an algorithm optimized specifically for use with TaqMan-based reagents, recommended primer⁄probe concentrations, and default thermal cycling conditions. Primer Express Software also supports assays based on the SYBR Green I dye. This minimizes the need for assay optimization and streamlines the design process.
The software also allows customized designs if the probe and⁄or one or more primers are known. Default and adjustable parameters (Figure 1) provide the flexibility required for all users.
Simple to Use
Simplicity and flexibility are the hallmarks of Primer Express Software v3.0.1, which is fully compatible with all StepOne, StepOnePlus, 7300, 7500, 7500 Fast, 7900HT, ViiA 7, and QuantStudio real-time PCR systems. Whether your research includes gene quantitation or allelic discrimination studies, Primer Express Software belongs in your lab.
Automated and Manual Primer⁄Probe Design
Primer Express Software v3.0.1 will design your primers and probes automatically, or you can design them manually. For automated design, simply choose either TaqMan or TaqMan MGB assays. If you know the sequence, you need only to import (or copy and paste) the sequence and quickly view your results. Alternatively, if you prefer to design the primers and probes manually, the procedure is equally simple and straight-forward as well.
Compatibility with Assay Development Guidelines
Primer Express Software v3.0.1 was designed to be compatible with standard guidelines for use of our real-time PCR instruments and reagents. These guidelines are such that traditional variables, including magnesium chloride concentration and the thermal cycling protocol, should be standardized by using a master mix of reagents (enzyme, salts, buffer) and tailored primer and probe design. This approach substantially reduces assay development time and improves laboratory efficiency by allowing multiple assays per plate to run simultaneously as all require the same conditions.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Real Time PCR (qPCR)
For Use With (Equipment)7300 System, 7500 Fast System, 7500 System, QuantStudio™ 12k Flex, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™, StepOnePlus™, ViiA™ 7 System
Operating SystemWindows 10
Product LinePrimer Express
Quantity5 users
Software TypePrimer Design
Product TypeLicense
Unit SizeEach
Contents & Storage
Store at Room Temperature, Included w/ StepOne, StepOnePlus, 7300, 7500, 7500 Fast, 7900HT, ViiA 7, QuantStudio 6, QuantStudio 7, and QuantStudio 12k Flex, Runs computers on Windows 10 OS for 7500, 7500 Fast, QuantStudio 6, QuantStudio 7, and QuantStudio 12K Flex instruments, Runs computers on Windows XP OS for StepOne, StepOnePlus, 7300, 7900HT, and ViiA 7 instruments, Runs on Windows 7 32-bit or Windows XP OS for non-instrument-controlling computers
Frequently asked questions (FAQs)
I need to see all the primer and probe sequences from an assay design right away. How can I do this?
You can directly see all the primer and probe sequences from a test design right away using the Primer Express Software. Open the software and choose File => New. Choose either TaqMan MGB Quantification or TaqMan Quantification for Gene Expression Assay design. Copy/paste your gene sequence of interest into the new window and click on the green triangle to get primer/probe designs.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Can I use Primer Express software on Windows 10?
Yes, you can use Primer Express v3.0.1 on Win10 computers.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
When designing primer and probes, what is difference between green amplicons and red amplicons in the "Map" display of Primer Express 2.0 software?
Primer and probe sets that satisfy the optimal length and Tm requirements are in red. Sets that don't satisfy one or the other or both of these parameters are in blue or green.
I am trying to install Primer Express Software, version 2.0 on my new computer with Windows XP as the operating system. It is giving me an error message, "Could not open Primers Express Preferences, because there is not enough memory" when trying to install or open the software despite this being a new computer. What can I do?
The Primer Express 2.0 software was developed to work on Windows 2000 operating systems. The "not enough memory" error message is generated due to incompatibility with the newer processor in the computers and the algorithm of the Primer Express v2.0 software. We have developed a patch to help over come this error message. You can download this patch from our website by going to the software downloads option under the Technical Resources page on our website. Click on ABI PRISM 7000 Sequence Detection System, then click on the PC link next to Primer Express P4 Enabler. Also download the installation instructions by clicking on the Read Me link.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
I get the following message when trying to start the Primer Express software: "Error opening Archive file: Primer Express is unable to open the archive file. Please move this file to another directory and run the Primer Express software again to create a new Archive." How do I solve this problem?
This error message can occur while attempting to install the Primer Express software, or sometimes when launching the software after it has been installed on the computer. To create a new archive file, hold down Alt key (to the left of the space bar) while opening the Primer Express application. You should get the browser window shown below. Click on “New” to create a New Archive.
The name “PXArchive” will automatically populate in the file name field. You may keep that name or change it to something different. Once you've named the Archive file, click on save. You should now be able to continue with the installation or use of the Primer Express Software.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.