(S)-1-BOC-3-hydroxypiperidine, 97%, Thermo Scientific Chemicals
(S)-1-BOC-3-hydroxypiperidine, 97%, Thermo Scientific Chemicals
(S)-1-BOC-3-hydroxypiperidine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(S)-1-BOC-3-hydroxypiperidine, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantityPackaging
4364400505 gGlass bottle
Catalog number 436440050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS143900-44-1
IUPAC Nametert-butyl 3-hydroxypiperidine-1-carboxylate
Molecular FormulaC10H19NO3
InChI KeyUIJXHKXIOCDSEB-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)N1CCCC(O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation+9° to +11° (20°C, 589 nm) (C=1 in chloroform)
Appearance (Form)Powder
GC>=96.0 %
Specific optical rotation+21° to +24° (20°C, 589 nm) (c=5 in MeOH)
Appearance (Color)White to tan
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only