TaqMan™ Fast Universal PCR Master Mix (2X), no AmpErase™ UNG, 500 reactions - FAQs
TaqMan™ Fast Universal PCR Master Mix (2X), no AmpErase™ UNG, 500 reactions - FAQs
View additional product information for TaqMan™ Fast Universal PCR Master Mix (2X), no AmpErase™ UNG - FAQs (4366073, 4366072, 4367846, 4364103, 4352042)
1 product FAQs found
Is the TaqMan Fast Universal PCR Master Mix offered with AmpErase UNG?
The TaqMan Fast Universal PCR Master Mix is provided without any UNG and you should add in AmpEraseUNG to reactions separately, with the addition of an initial 2 minute cycling step at 50º C. If you prefer to have a master mix with the UNG pre-added, please use TaqMan Fast Advanced Master Mix.