What is the target specificity of the TaqMan Campylobacter jejuni Detection Kit (P/N 4366098)?
The kit demonstrates 100% inclusivity for 27 strains of Campylobacter jejuni and Campylobacter coli and 100% exclusivity for 29 other Campylobacter jejuni and Campylobacter coli strains. For more specific information, please refer to “Appendix A: Specificity” of the TaqMan Campylobacter jejuni Detection Kit Protocol. Also, included there is the American Tissue Culture and Collection Strains / Source information.