TaqMan® E. coli O157:H7 Detection Kit - FAQs
TaqMan® E. coli O157:H7 Detection Kit - FAQs
View additional product information for TaqMan® E. coli O157:H7 Detection Kit - FAQs (4366100)
2 product FAQs found
What is the target specificity of the TaqMan Escherichia coli O157:H7 Detection Kit (P/N 4366100)?
The TaqMan Escherichia coli O157:H7 Detection Kit has demonstrated 100% inclusivity for 16 strains of serotypes 0157:H7 and 055:H7. It has also demonstrated 100% exclusivity against 31 other Escherichia coli strains and 21 non-Escherichia coli strains. For more specific information, please refer to “Appendix A: Specificity” of the TaqMan E. coli 0157:H7 Detection Kit Protocol. Also, included there is the American Tissue Culture and Collection Strains / Source information.
Why are each of the individual components in the TaqMan Pathogen Detections Kits color-coded?
Each individual component in the TaqMan Pathogen Detection Kits is color-coded in order to best distinguish between the 2X Environmental Master Mix, the negative control and the 10X Target Assay Mix (TAM). Yellow coded vials are 2X Environmental Master Mix, the white coded vial is the negative control solution, and the 10X Target Assay Mix (TAM) which is associated with a specific organism is a gray-labeled vial.