1-BOC-4-(Aminomethyl)piperidine, 97%
1-BOC-4-(Aminomethyl)piperidine, 97%
1-BOC-4-(Aminomethyl)piperidine, 97%
Thermo Scientific Chemicals

1-BOC-4-(Aminomethyl)piperidine, 97%

CAS: 144222-22-0 | C11H23N2O2 | 215.32 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4366100505 gGlass bottle
Catalog number 436610050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS144222-22-0
IUPAC Name{1-[(tert-butoxy)carbonyl]piperidin-4-yl}methanaminium
Molecular FormulaC11H23N2O2
InChI KeyKLKBCNDBOVRQIJ-UHFFFAOYSA-O
SMILESCC(C)(C)OC(=O)N1CCC(C[NH3+])CC1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Color)Colorless to yellow
Appearance (Form)Liquid or low melting solid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only