Product Image
Applied Biosystems™
GeneMapper™ Software v4.1 Documentation only
The GeneMapper™ Software v4.1 printed documentation includes the Installation and Administration Guide, Reference and Troubleshooting Guide, Installation Options User Bulletin,Read more
Have Questions?
Catalog NumberQuantity
43668531 set
Catalog number 4366853
Price (USD)
646.00
1 set
Quantity:
1 set
Price (USD)
646.00
1 set
The GeneMapper™ Software v4.1 printed documentation includes the Installation and Administration Guide, Reference and Troubleshooting Guide, Installation Options User Bulletin, Autoanalysis User Bulletin, Quick Reference Guide and Getting Started Guides that describe different workflows (Microsatellite, SNapShot™ Kit, SNPlex™ System, AFLP and LOH analysis), as well as a documentation CD.
GeneMapper™ Software is a flexible genotyping software package that provides DNA sizing and quality allele calls for all of our electrophoresis-based genotyping systems. This software specializes in multiapplication functionality, including amplified fragment length polymorphism (AFLP), loss of heterozygosity (LOH), microsatellite, and SNP genotyping analysis. GeneMapper™ Software can help users increase data processing efficiency with remote auto-analysis and command line operation, and allows for multiuser, client-server deployment. The software uses Process Quality Values (PQVs) for automated identification that reduces data review time for high throughput genotyping. In addition, the security and audit features help users meet 21 CFR 11 requirements.
GeneMapper™ Software new features include:
• Co-installation and autoanalysis with the 3500 series systems Data Collection v1.0 Software on Windows Vista™ 32-bit Business operating system, Service Pack 1.
• Co-installation and autoanalysis with the 3130 and 3730 series systems Data Collection v3.1 Software on Microsoft Windows™ XP Professional (Service Pack 2 or 3) or Vista™ 32-bit Business (Service Pack 1) operating systems to save time in the analysis pipeline.
• A new Oracle™ 11g database with project-related data storage ensures faster installation times and latest database support, as well as flexible archive capabilities through the utilities provided with the GeneMapper™ software.
• Users will be able to customize the project auto-saving frequency in the Options menu.
• Users can now define a linearity range in the analysis methods for more reproducible, higher quality sizing of larger fragments (>800bps).
• Analysis of 6-dye data generated from the Applied Biosystems 3500 and 3500xL instrument systems for higher multiplexing capabilities.
• Many user interface and feature improvements.
GeneMapper™ Software has not undergone specific developmental validation for human identification applications. Human identification laboratories analyzing single-source or parentage samples which choose to use GeneMapper™ Software for data analysis should perform their own developmental validation studies.
Several users may access the database concurrently; however, because genotype analysis is data-intensive, more than five clients simultaneously accessing the database may slow the performance of GeneMapper software.
For Research Use Only. Not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Sequencing
Product LineGeneMapper
Product TypeSoftware
Quantity1 set
Unit Size1 set
Frequently asked questions (FAQs)
How can I import Project Information into GeneMapper Manager?
Go to the Tools menu and select GeneMapper Manager. Click on the tab for the file(s) you want to import. Click on Import at the bottom of the window and select the files to be imported. Note: Multiple projects can be selected for importing at one time, but all other tabs are one at a time. Once imported, the new files will appear in the list. When finished, click Done in the lower right corner.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
How can I back up my data files for DNA sequencing and fragment analysis applications, with the following extension, .ab1 or .fsa, on the capillary electrophoresis instruments?
The location of the sample files, .ab1 or .fsa, will either be in the Runs folder (D:/AppliedBio/310/Runs) on the ABI PRISM 310 Genetic Analyzer system or it will be determined by the Results Group in the Data Collection Software on the multi-capillary platforms. If you do not know where that location is, you can go to the Data Collection Software for your instrument and go to the Results Group section (top left corner in Unified Data Collection; for 3500 Data Collection, go to Library and in the Manage section, select Results Group), open the Results Group, and check the Sample File Folder location.
Save the files to the storage media of your choice (CD, DVD, network drive, flash drive).
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Do you have a manual uninstall procedure for Full Install configurations on a stand-alone computer for GeneMapper Software v.4.1?
Please download this PDF (http://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/GeneMapper-Software-v.4-uninstall-full-install.pdf) and follow the directions to uninstall GeneMapper Software v.4.1.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
I need to uninstall GeneMapper Software v.4.1 Client because of failed installation. How can I do this for Client Install configurations on a stand-alone computer?
Please download this PDF (http://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/GeneMapper-Software-v.4-1-uninstall-client-install.pdf) and follow the directions to uninstall GeneMapper Software v.4.1.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
We are upgrading to the Windows 7 operating system. Can I install GeneMapper Software v.4.1 on that?
Unfortunately, GeneMapper Software v.4.1 is not compatible with Windows 7. Please click here (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/GeneMapper-Software-v.4-1-datasheet.pdf) for the GeneMapper Software v.4.1 datasheet.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.