Product Image
Applied Biosystems™

Precision Plate Holder for 0.2 mL Tubes & Strips (for 7300 & 7500 Systems)

This precision plate holder is for use with 0.2 mL tube strips on the Applied Biosystems™ 7300 and 7500 Real-Time PCR systems.
Have Questions?
Catalog NumberQuantity
43670330.2 mL
Catalog number 4367033
Price (USD)
706.00
Each
Quantity:
0.2 mL
Price (USD)
706.00
Each

This precision plate holder is for use with 0.2 mL tube strips on the Applied Biosystems™ 7300 and 7500 Real-Time PCR systems.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Application)Real Time PCR (qPCR)
For Use With (Equipment)7300 System, 7500 System
FormatTube
PCR MethodqPCR
Plate Compatibility8-well Strips
Quantity0.2 mL
Reaction SpeedFast
Shipping ConditionRoom Temperature
Product TypePrecision Plate Holder
Unit SizeEach
Contents & Storage
Store at Room Temperature