Ammonium dihydrogen phosphate, 99.999%, (trace metal basis)
Ammonium dihydrogen phosphate, 99.999%, (trace metal basis)
Ammonium dihydrogen phosphate, 99.999%, (trace metal basis)
Thermo Scientific Chemicals

Ammonium dihydrogen phosphate, 99.999%, (trace metal basis)

CAS: 7722-76-1 | H6NO4P | 115.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number 436791000
Price (USD)
155.65
Online Exclusive
173.00
Save 17.35 (10%)
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
155.65
Online Exclusive
173.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS7722-76-1
IUPAC Namephosphoric acid amine
Molecular FormulaH6NO4P
InChI KeyLFVGISIMTYGQHF-UHFFFAOYSA-N
SMILESN.OP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Total trace metal impurities=<10 ppm
Appearance (Color)Colorless to white
Titration with NaOH>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only