6-Fluoropyridine-2-carboxylic acid, 97%
6-Fluoropyridine-2-carboxylic acid, 97%
6-Fluoropyridine-2-carboxylic acid, 97%
Thermo Scientific Chemicals

6-Fluoropyridine-2-carboxylic acid, 97%

CAS: 402-69-7 | C6H4FNO2 | 141.10 g/mol
Have Questions?
Catalog NumberQuantity
4368400101 g
Catalog number 436840010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS402-69-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Form)Crystalline powder
Melting point135°C to 142°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only