2-Bromo-5-iodopyridine, 97%
2-Bromo-5-iodopyridine, 97%
2-Bromo-5-iodopyridine, 97%
Thermo Scientific Chemicals

2-Bromo-5-iodopyridine, 97%

CAS: 73290-22-9 | C5H3BrIN | 283.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 436850050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS73290-22-9
IUPAC Name2-bromo-5-iodopyridine
Molecular FormulaC5H3BrIN
InChI KeyLLKRSJVPTKFSLS-UHFFFAOYSA-N
SMILESBrC1=CC=C(I)C=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to light pink to light brown
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Crystalline powder or needles
Melting point121°C to 125°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only