TaqMan™ Staphylococcus aureus Detection Kit - FAQs
TaqMan™ Staphylococcus aureus Detection Kit - FAQs
View additional product information for TaqMan™ Staphylococcus aureus Detection Kit - FAQs (4368606)
1 product FAQs found
Why are each of the individual components in the TaqMan Pathogen Detections Kits color-coded?
Each individual component in the TaqMan Pathogen Detection Kits is color-coded in order to best distinguish between the 2X Environmental Master Mix, the negative control and the 10X Target Assay Mix (TAM). Yellow coded vials are 2X Environmental Master Mix, the white coded vial is the negative control solution, and the 10X Target Assay Mix (TAM) which is associated with a specific organism is a gray-labeled vial.