TaqMan® Salmonella enterica Detection Kit & PrepMan® Ultra Reagent - FAQs
TaqMan® Salmonella enterica Detection Kit & PrepMan® Ultra Reagent - FAQs
View additional product information for TaqMan® Salmonella enterica Detection Kit & PrepMan® Ultra Reagent - FAQs (4368723)
3 product FAQs found
Where is the DNA layer after using the PrepMan Ultra Sample Preparation Reagent (P/N 4318930)?
After the final step of centrifugation, the DNA is in the supernatant. The supernatant should not be discarded. The pellet contains protein and other cellular components that are digested during the 10 minute incubation of the lysate at 100 deg C. For more information please refer to the PrepMan Ultra Sample Prep Reagent Protocol.
Why is it important to dilute the final supernatant when using PrepMan Ultra Sample Preparation Reagent?
The final supernatant must be diluted at least 1:10 to minimize the amount of potential inhibitors from the sample and from the PrepMan Ultra Sample Preparation Reagent (P/N 4318930). In some cases, highly concentrated cultures (at about 10E8 cfu/ml or more) or residual PrepMan Ultra reagent can form gel-like masses that may inhibit PCR.
What is the purpose of performing pre-enrichment on food samples for isolation of DNA with PrepMan Ultra reagents?
Without performing any specific pre-enrichment step on certain food samples like meat, the use of the PrepMan Ultra Sample Preparation Reagent (P/N 4318930) will not enable the reduction of many forms of PCR inhibitors such as fats, tissues and particulates. Thus, pre-enrichment significantly reduces the detection of false positive signal from dead or nonpathogenic cells in food samples. Improper pre-enrichment steps can hamper the process of extracting DNA of the highest purity and affect the performance of TaqMan Pathogen Detections kits for Salmonella enterica, Listeria monocytogenes, Campylobacter jejuni, E. coli 0157:H7, or MicroSeq kits.