Which kit should I use for the reverse transcription step for TaqMan Pri-miRNA assays (Cat. No. 4427012)?
We recommend the following kits:
Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.
- High-Capacity cDNA Reverse Transcription Kit (Cat. No. 4374966).
- High-Capacity RNA-to-cDNA Kit (Cat. No. 4387406).
Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.