Potassium bromide, 99.999%, (trace metal basis)
Potassium bromide, 99.999%, (trace metal basis)
Potassium bromide, 99.999%, (trace metal basis)
Acros Organics

Potassium bromide, 99.999%, (trace metal basis)

CAS: 7758-02-3 | BrK | 119.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Plastic bottle
Catalog number 436950100
also known as 43695
HSN 28275120
Price (INR)
12,933.00
Each
Quantity:
10 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
12,933.00
Each
Chemical Identifiers
CAS7758-02-3
IUPAC Namepotassium bromide
Molecular FormulaBrK
InChI KeyIOLCXVTUBQKXJR-UHFFFAOYSA-M
SMILES[K+].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Titration Argentometric33 % (K) Typical
Total trace metal impurities=<10 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only