Tetraammineplatinum(II) chloride hydrate, 99.995%, (trace metal basis)
Tetraammineplatinum(II) chloride hydrate, 99.995%, (trace metal basis)
Tetraammineplatinum(II) chloride hydrate, 99.995%, (trace metal basis)
Tetraammineplatinum(II) chloride hydrate, 99.995%, (trace metal basis)
Thermo Scientific Chemicals

Tetraammineplatinum(II) chloride hydrate, 99.995%, (trace metal basis)

CAS: 108374-32-9 | Cl2H12N4Pt | 334.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
5 g
Packaging:
Glass bottle
Catalog number 437000050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS108374-32-9
IUPAC Nameplatinum(2+) tetraamine dichloride
Molecular FormulaCl2H12N4Pt
InChI KeyKHCPSOMSJYAQSY-UHFFFAOYSA-L
SMILESN.N.N.N.[Cl-].[Cl-].[Pt++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total trace metal impurities=<50 ppm
Appearance (Color)Off-white to light yellow
Appearance (Form)Crystalline powder
Platinum (Pt)51 to 59 % (typical)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only