Not what you're looking for? Search all of our TaqMan® arrays for your gene or pathway of interest.
Do you need this panel in a 384-well plate? Please visit Specialty Plating to learn more.
TaqMan™ Array Human Immune Panel
Applied Biosystems™

TaqMan™ Array Human Immune Panel

The TaqMan™ Human Immune Array from Applied Biosystems™ is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysisRead more
Have Questions?
Catalog NumberQuantity
43705734 cards
Catalog number 4370573
Price (USD)
2,645.00
Each
Quantity:
4 cards
Price (USD)
2,645.00
Each
The TaqMan™ Human Immune Array from Applied Biosystems™ is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of targets known to have implications in immune response. This product is designed with 96 TaqMan™ Gene Expression Assays loaded in singletons into the array’s 384 wells and is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System. The targets include cytokines, chemokines, growth factors, immune regulators, apoptosis markers, ischemia markers, tissue-specific markers, and others including classic endogenous markers.This product is sold in 1 box of 4 cards.

The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormDried Down
IncludesTaqMan Assay for One 2 uL Reaction
Internal Probe ModificationMGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
Label or DyeFAM
Product LineTaqMan
Quantity4 cards
Reaction SpeedStandard
Research CategoryCell Cycle & Proliferation, Apoptosis, Neurobiology, Signal Transduction, Biomarker-Related Pathway(s), Extracellular Matrix & Cell Adhesion, Reproduction, Toxicology & Drug Metabolism, Immunology, Oncology
Target GeneCytokines, chemokines, growth factors, and other immune response genes
TypeArray Human Immune Panel
Format384-well Microfluidic Card
SpeciesHuman
Unit SizeEach
Contents & Storage
Each TaqMan™ Array well contains a single TaqMan™ Assay for one 2 μL reaction. The cards should be stored at 4°C.

Frequently asked questions (FAQs)

What are the storage recommendations for TaqMan Array Cards (TAC)?

We recommend storing TaqMan Array Cards (TAC) at 2-8 degrees C. TaqMan Array Cards (TAC) are stable for 2 years from the manufacturing date. The expiration date is listed on the packaging.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.