SurePac™ Oligo RP MDi™ HPLC Columns
Thermo Scientific™

SurePac™ Oligo RP MDi™ HPLC Columns

SurePac Oligo RP MDI is a robust reversed-phase column designed for oligonucleotide analysis by HPLC and LC-MS. It delivers high recovery, bioinert column hardware, and reproducible separations, enabling reliable profiling of oligonucleotides of any size.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Length (Metric)
43712-051003210 mm50 mm
43712-0221322.1 mm20 mm
43712-0521322.1 mm50 mm
43712-1021322.1 mm100 mm
43712-050332300 μm50 mm
43712-052123221.2 mm50 mm
Catalog number 43712-0510032
Price (USD)
-
Diameter (Metric):
10 mm
Length (Metric):
50 mm
Thermo Scientific™ SurePac™ Oligo RP MDi™ HPLC columns deliver high-resolution, reproducible separations for therapeutic oligonucleotides and nucleic acids across R and D and QC workflows. Built on monodisperse, supermacroporous polymeric packing with bio-inert hardware, they provide stable performance even under elevated pH and temperature conditions, resolving truncation impurities while supporting large oligos and intact nucleic acids for scalable LC-MS and LC-UV analysis.
  • Purpose-built for oligonucleotide RP separations
  • Optimized for DNA and RNA oligonucleotides, including modified sequences, delivering consistent retention and selectivity across short and long oligos.
  • Bioinert, metal-free fluid path
  • Reduces metal/oligonucleotide interactions to improve recovery, minimize adsorption, and deliver cleaner peak shapes for sensitive analyses.
  • Optimized for ion-pair RP LC-MS
  • Fully compatible with common ion-pairing reagents such as HFIP/TEA, enabling sensitive, reproducible LC-MS detection.
  • High recovery of long and modified oligonucleotides
  • High resolution power for oligonucleotides spanning over a wide size range from small to (ultra) large
  • Supports efficient analysis of long, hydrophobic, and chemically complex oligos without compromising signal or resolution.
  • Robust stationary phase for demanding methods
  • Maintains stable performance under high organic content and elevated temperatures, supporting fast gradients and long column lifetimes.
  • Excellent peak shape and impurity resolution
  • Delivers sharp, symmetrical peaks and reliable separation of closely related species, including impurities and sequence variants.
  • Reproducible, manufacturing-controlled performance
  • Ensures consistent results across columns and batches, simplifying method transfer and routine use.
  • Flexible for development and routine workflows
  • Suitable for method development, impurity profiling, and QC, reducing the need for multiple column chemistries.
  • Compatible with UHPLC and HPLC systems
  • Designed for seamless integration into modern LC platforms and existing laboratory setups.
  • Built for confident bioanalytical results
  • Improves robustness, reduces variability, and supports reliable data generation in oligonucleotide workflows.
Specifications
Column FormatAnalytical
Flow Rate1 to 5 mL/min.
Max. Pressure5600 psi (380 bar)
Packing MaterialPolymeric
Stationary PhasePoly-DVB
Temperature5°C to 90°C
Diameter (Metric)10 mm
Length (Metric)50 mm
Particle Size2.5 μm
Pore SizeProprietary supermacroporous
pH2 to 12
Unit SizeEach