Product Image
Applied Biosystems™
MicroSEQ™ ID 16S rDNA Full Gene Library v2.0
The MicroSEQ™ ID 16S rDNA Full Gene Library v2.0 is designed for installation with the MicroSEQ™ ID analysis software.
Have Questions?
Catalog NumberQuantity
43712951 Kit
Catalog number 4371295
Price (MXN)
-
Quantity:
1 Kit
The MicroSEQ™ ID 16S rDNA Full Gene Library v2.0 is designed for installation with the MicroSEQ™ ID analysis software. It contains the complete 16S sequence from bacterial organisms that can be compared to an unknown bacterial sequence using MicroSEQ™ ID analysis software to identify bacteria.
MicroSEQ™ ID Analysis Software enables you to identify and classify unidentified bacterial or fungal sequences by comparing them to a validated microbial library.
  • A validated library of over 2300 microbial sequences ensures you get the answers you need.
  • Easy-to-use interface and optimized search algorithms enable quick results.
  • Phylogenetic trees provide a deeper understanding of your unknowns.
  • Security features support 21 CFR Part 11 compliance, enabling the software to integrate smoothly into your workflow.
Confidently Identify Microbes
The MicroSEQ™ system comprises bacterial libraries of 500 bp sequences and of over 1,400 validated entries--including extensive coverage of gram-negative nonfermenters, Bacillus, Coryneforms, Mycobacteria, and Staphylococcus. Another library containing more than 900 entries is available for fungal identification. To ensure you can accurately identify the species of your sequences, we rigorously validate all library entries. And unlike other widely available databases, MicroSEQ libraries contain only correctly identified organisms and quality-controlled sequences derived from type strains of preeminent culture collections. The libraries also support sequence polymorphisms, and include bacterial Category A biothreat agents.
Flexible and Easy-to-Use
To simplify microbial sequence identification, analysis, and reporting, MicroSEQ ID Analysis Software provides easy-to-use wizards and a powerful, automated toolset. The software also includes a full complement of audit-trail and security features that support 21 CFR Part 11 compliance. Further, this flexible software even allows you to create customized entries for specific sequences from new or proprietary strains.
The MicroSEQ™ System: A Complete, Integrated Solution
Streamlining every step in microbial identification, Applied Biosystems MicroSEQ System combines the advantages of MicroSEQ ID Software with easy PrepMan Ultra sample preparation reagents and protocols, MicroSEQ bacterial and fungal application kits, and industry-leading thermal cyclers and sequencing systems. To ensure optimal performance and system-wide integration, we develop and test all components of the system together.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Sequencing
Product LineMicroSEQ
Product TyperDNA Full Gene Library
Quantity1 Kit
Unit SizeEach
Frequently asked questions (FAQs)
You offer MicroSEQ ID 16S rDNA Full Gene Library v2.0, USB format (Cat. No. A46365) and MicroSEQ ID 16S rDNA Full Gene Library v2.0 (Cat. No. 4371295). What is the difference between them?
The only difference is that Cat. No. A46365 is available in USB format, otherwise the content is the same between the two catalog numbers.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What is the difference in library format between MicroSEQ ID 16S rDNA Full Gene Library v2.0 and MicroSEQ ID 16S rDNA Full Gene Library v1.0?
MicroSEQ ID 16S rDNA Full Gene Library v2.0 is the same as MicroSEQ ID 16S rDNA Full Gene Library 1.0 but in USB format. No new content has been added.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.