NA-<i>Star</i>&trade; Influenza Neuraminidase Inhibitor Resistance Detection Reagent Set
Invitrogen™

NA-Star™ Influenza Neuraminidase Inhibitor Resistance Detection Reagent Set

Designed for rapid and sensitive quantitation of influenza neuraminidase inhibitor resistance in dilutions of virus culture medium in a 96-well microplate format. Thermo Scientific NA-Star Influenza Neuraminidase Inhibitor Reagent kit provides all necessa

Have Questions?
Catalog NumberQuantity
43743481 set
Catalog number 4374348
Price (USD)
2,030.00
Each
Quantity:
1 set
Request bulk or custom format
Price (USD)
2,030.00
Each

The NA-Star™ Influenza Neuraminidase Inhibitor Resistance Detection Reagent Set is designed for the rapid and sensitive quantitation of influenza neuraminidase inhibitor resistance in dilutions of virus culture medium in a 96-well microplate format. The NA-Star™ Influenza Neuraminidase Inhibitor Reagent kit provides all necessary assay reagents enabling improved global assay standardization and more accurate comparison of results lab-to-lab.

The NA-Star™ Influenza Neuraminidase Inhibitor Resistance Detection Kit includes everything you need to quantitate neuraminidase activity and neuraminidase inhibitor resistance in avian, equine, human (types A and B), and porcine influenza viruses. The kit's fast and easy protocol and convenient 96-well plate format make it ideal for monitoring influenza virus neuraminidase inhibitor resistance, as well as high-throughput inhibitor compound screening.

  • Includes pre-made solutions for all required reagents, enabling you to standardize assays and compare results between labs easily.
  • Provides up to 50-fold higher sensitivity than assays using fluorescent methylumbelliferone N-acetylneuraminic acid (MUNANA) substrate.
  • Dynamic range of four orders of magnitude enables you to quantitate virus isolates over a broad range of virus concentration and with varying levels of neuraminidase activity without performing numerous sample dilutions.
  • Fast and easy protocol enables you to complete assays in less than 1.5 hours, providing rapid assay throughput.

Get Results in Less than 1.5 Hours

The fast and easy protocol enables you to perform assays in less than 1.5 hours. Simply incubate your virus samples with dilutions of neuraminidase inhibitor, add NA-Star chemiluminescent substrate, incubate again, and then add the accelerator solution, which triggers light emission from the reaction product. Light signal is measured with a luminometer, including multi-mode instruments that include a luminometer mode. For best results, use a luminometer with an automatic injector to add the accelerator solution. You may use a luminometer without an injector, provided you add reagents with a multichannel pipettor and read the plate immediately.

Up to 50-Fold Higher Sensitivity than MUNANA-Based Assays

The chemiluminescent-based detection technology provides a wide dynamic range -- greater than four orders of magnitude of neuraminidase concentration (two orders of magnitude greater than fluorescent MUNANA-based assays) -- enabling you to quantitate neuraminidase inhibitor resistance levels over a broad range of virus concentration and neuraminidase activity without having to test multiple virus dilutions.

Pre-Made Reagents Make it Easy to Standardize Assays

Quality-tested protocols, and pre-made reagents eliminate variability, enabling you to easily standardize assay performance and results across experiments and different laboratories.

Monitor Drug Resistance of Human, Avian, and Livestock Influenza Viruses

The kit's ease of use, standardization, and broad range of compatible species (avian, equine, human types A and B, and porcine influenza viruses) make it an important new tool for assessing and researching drug resistance of influenza in humans, birds, and livestock.

Screen for New Inhibitors and Develop New Vaccines

The kit has many other applications, including screening for new neuraminidase inhibitors and quantitating viral neuraminidase activity for vaccine development. It is also ideal for quantitating neuraminidases from bacteria, including S. typhimurium, C. perfringens, V. cholera, and likely others.

For Research Use Only. Not for use in diagnostic procedures.

WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodLuminescence
FormLiquid
Format96-well plate
Quantity1 set
Shipping ConditionWet Ice
SubstrateNA-Star™
Substrate PropertiesChemical Substrate
Substrate TypeNeuraminidase Substrate
Target EnzymeSialidase, Neuraminidase
ThroughputHigh-throughput Compatible
Validated ApplicationEnzyme Assay
For Use With (Application)Neuraminidase (Sialidase) Assay, Inhibitor Resistance Detection
For Use With (Equipment)Luminometer
Product LineNA-Star, NovaBright
Product TypeNeuraminidase Assay
Reference Wavelength600 nm
Unit SizeEach
Contents & Storage
The set includes the substrate, required buffer, and reaction accelerator. Store all components at 2°C to 8°C.