Sodium hexanoate, 99%
Sodium hexanoate, 99%
Sodium hexanoate, 99%
Thermo Scientific Chemicals

Sodium hexanoate, 99%

CAS: 10051-44-2 ∣ C6H11NaO2 ∣ 138.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
Catalog number 437540100
Price (USD)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS10051-44-2
IUPAC Namesodium hexanoate
Molecular FormulaC6H11NaO2
InChI KeyUDWXLZLRRVQONG-UHFFFAOYSA-M
SMILES[Na+].CCCCCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Infrared spectrumConforms
Loss on drying=<2 %
Titration with HClO4>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only