3-Fluorobenzyl isocyanate, 98%
3-Fluorobenzyl isocyanate, 98%
3-Fluorobenzyl isocyanate, 98%
Thermo Scientific Chemicals

3-Fluorobenzyl isocyanate, 98%

CAS: 102422-56-0 | C8H6FNO | 151.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 437580050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS102422-56-0
IUPAC Name1-fluoro-3-(isocyanatomethyl)benzene
Molecular FormulaC8H6FNO
InChI KeyPHRJTGPFEAUEBC-UHFFFAOYSA-N
SMILESFC1=CC=CC(CN=C=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only