The Applied Biosystems™ MicroAmp™ Fast Optical 48-Well Reaction Plate is is optimized to provide unmatched temperature accuracy and uniformity forLeia mais
Have Questions?
Número do catálogo
Quantity
4375816
20 Plates
Número do catálogo 4375816
Preço (BRL)
618,57
Each
Quantity:
20 Plates
Preço (BRL)
618,57
Each
The Applied Biosystems™ MicroAmp™ Fast Optical 48-Well Reaction Plate is is optimized to provide unmatched temperature accuracy and uniformity for fast, efficient PCR amplification. This plate, constructed from a single rigid piece of polypropylene in a 48-well format, is compatible with the Applied Biosystems™ StepOne™ Real-Time PCR System.
• Maximum thermal conductivity for precise thermal cycling • Reproducible, specific, and sensitive real-time PCR results in just 40 minutes • Validated with other system components for consistent results • Each reaction plate includes a unique serialized, eight character number label (barcode) that is user-readable and machine-readable to prevent tracking errors
Increased Thermal Contact for Faster, More Uniform Heating The advanced design of the MicroAmp™ Fast 48-Well Reaction Plate dramatically increases its thermal contact with the Applied Biosystems™ StepOne™ Real-Time PCR System. The plate also has a consistent well-to-well thickness so all of the samples cycle through the PCR process uniformly. These features reduce experimental artifacts and help ensure fast, accurate results.
Especificações
Certifications/ComplianceCertified DNA/RNase Free
For Use With (Equipment)Thermal Cycler, Genetic Analyzer, DNA Analyzer
Packaging20 Plates
Product TypeMicroplate
Quantity20 Plates
Reaction SpeedFast
Shipping ConditionRoom Temperature
Skirt StyleSemi
Volume (Metric) Well0.2 mL
Volume (Metric) Working0.1 mL
BarcodeNo Barcode
ColorOptical
MaterialPolypropylene
No. of Wells48
Product LineMicroAmp
Unit SizeEach
Conteúdo e armazenamento
Contains 20 plates.
May be stored at room temperature. Under proper storage conditions, plates may be stored indefinitely.
Frequently asked questions (FAQs)
Can I cut a 96-well FAST plate to fit the StepOne system?
No, the MicroAmp Fast Optical 48-Well Reaction Plate (P/N 4375816) is the only plate that can be used on the StepOne system. Using a plate that has not been designed for use with the StepOne Real-Time PCR System is not recommended and can potentially damage the instrument.