MicroAmp™ 48-Well Optical Adhesive Film, 25 films - FAQs
MicroAmp™ 48-Well Optical Adhesive Film, 25 films - FAQs
View additional product information for MicroAmp™ 48-Well Optical Adhesive Film - FAQs (4375928, 4375323)
2 product FAQs found
Can I cut an Optical Adhesive seal in half to fit the 48well StepOne system plate?
It is not recommended that you do so. Please use the MicroAmp 48-Well Optical Adhesive Film (P/N 4375928).
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Can the MicroAmp Optical Adhesive Film be used with MicroAmp Optical Tubes?
No. The Optical Adhesive Film is recommended for use only with the MicroAmp Optical 96-well Reaction Plate (with or without barcode) and MicroAmp 384-well Optical Reaction Plate.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.