2-Amino-3-methoxybenzoic acid, 98%
2-Amino-3-methoxybenzoic acid, 98%
2-Amino-3-methoxybenzoic acid, 98%
Acros Organics

2-Amino-3-methoxybenzoic acid, 98%

CAS: 3177-80-8 | C8H9NO3 | 167.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 437720010
also known as 43772
HSN 32049000
Price (INR)
7,175.00
Each
Quantity:
1 g
Request bulk or custom format
Price (INR)
7,175.00
Each
Chemical Identifiers
CAS3177-80-8
IUPAC Name2-amino-3-methoxybenzoic acid
Molecular FormulaC8H9NO3
InChI KeySXOPCLUOUFQBJV-UHFFFAOYSA-N
SMILESCOC1=CC=CC(C(O)=O)=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
HPLC>=97.5 %
Appearance (Color)White to yellow to brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only