VeriFlex™ 96-well Temperature Verification Kit for 0.2 mL Veriti™ System, 9 channel - FAQs
VeriFlex™ 96-well Temperature Verification Kit for 0.2 mL Veriti™ System, 9 channel - FAQs
View additional product information for VeriFlex™ 96-well Temperature Verification Kit for 0.2 mL Veriti™ System, 9 channel - FAQs (4377669)
2 product FAQs found
Do you offer an alternative for the discontinued Multiprobe Temperature Verification Kit (Cat. No. 4377669)?
No, we do not offer an alternative for this product.
Do you offer a temperature verification service to verify my PCR/qPCR instrument performance?
Yes, we offer an on-site temperature verification service with additional features such as ramp rate testing. Please find more information regarding the service below:
https://www.thermofisher.com/us/en/home/products-and-services/services/instrument-qualification-services/compliance-and-validation/on-site-temperature-verification.html